Learn More
Manganese(II) chloride tetrahydrate, 99.99% (metals basis)
Description
It is used as a source of manganese in biological research, in the production of dry cell batteries, and precursor to the antiknock compound. It is used in industrial applications such as dyeing disinfecting, the purification of natural gas, and as a drying agent for linseed oil. It is used in pigment manufacturing, fertilizers and in construction materials and also used in aluminum smelting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 58°C |
| Physical Form | Crystalline |
| Quantity | 25 g |
| Merck Index | 14,5728 |
| Solubility Information | Soluble in water,acetic acid,pyridine,and ethanol but insoluble in ether. |
| Formula Weight | 197.90 (125.84 Anhydrous) |
| Percent Purity | 99.99% |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 2.010 g/mL |
| Show More |
Safety and Handling
- Manganese(II) chloride tetrahydrate
- Danger
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H301-Toxic if swallowed.
- H318-Causes serious eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contain: Manganese chloride (MnCl2), tetrahydrate
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.