missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) chloride, 97%, Thermo Scientific Chemicals
62.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
25 g
250 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1190.0°C |
Melting Point | 650.0°C |
Color | Pink |
Physical Form | Flakes |
Quantity | 2.5 kg |
Assay Percent Range | 96% min. (Complexometry) |
Linear Formula | MnCl2 |
Fieser | 07,222 |
Merck Index | 15, 5793 |
Solubility Information | Solubility in water: 723g/L (25°C). Other solubilities: soluble in alcohol,insoluble in ether |
Show More |
Safety and Handling
Product Identifier
- Manganese(II) chloride
- Danger
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
- P270-Do not eat, drink or smoke when using this product.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'