missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) carbonate, 99.9% (metals basis excluding Na)

Product Code. 11307907
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307907 1 kg 1kg
11317907 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307907 Supplier Thermo Scientific Alfa Aesar Supplier No. 014344.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-62-9 | CMnO3 | 114.946 g/mol

Manganese(II) carbonate is used as an additive to plant fertilizers, glaze colorant in ceramics, pigment and drier for varnishes. It is also used as an ingredient in welding rod, animal feed additive as well as to prepare other manganese salts and pharmaceuticals. It is also used in medicine as a hematinic and in health foods, in flux and in concrete stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-62-9
Molecular Formula CMnO3
Molecular Weight (g/mol) 114.946
MDL Number MFCD00011116
InChI Key XMWCXZJXESXBBY-UHFFFAOYSA-L
Synonym manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
PubChem CID 11726
IUPAC Name manganese(2+);carbonate
SMILES C(=O)([O-])[O-].[Mn+2]

Specifications

Melting Point 350°C (decomposition)
Color Brown to Violet
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.9% (metals basis excluding Na)
Solubility Information Soluble in dilute inorganic acids and carbon dioxide. Insoluble in water,alcohol,ammonia and common organic acids.
Formula Weight 114.95
Odor Odorless
Appearance Light brown to violet powder
Density 3.125
Chemical Name or Material Manganese(II) carbonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.