missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide hydrate, 99.9% (metals basis)

Product Code. 11410750
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410750 10 g 10g
11420750 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410750 Supplier Thermo Scientific Alfa Aesar Supplier No. 044443.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10031-20-6 | Br2H8MnO4 | 286.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10031-20-6
Molecular Formula Br2H8MnO4
Molecular Weight (g/mol) 286.81
MDL Number MFCD00149791
InChI Key HHDPJRSXPOGIOP-UHFFFAOYSA-L
Synonym manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
IUPAC Name manganese(2+) tetrahydrate dibromide
SMILES O.O.O.O.[Mn++].[Br-].[Br-]

Specifications

Melting Point 64.3°C (decomposition)
Color Pink
Physical Form Crystalline
Quantity 10 g
Merck Index 14,5725
Formula Weight 214.76 (Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Manganese (II) bromide hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.