missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) bromide, 99%, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Pink |
Physical Form | Powder |
Quantity | 100 g |
Assay Percent Range | 99% |
Linear Formula | MnBr2 |
Merck Index | 15, 5790 |
Solubility Information | Solubility in water: soluble |
Formula Weight | 214.76 |
Percent Purity | 99% |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Manganese(II) bromide, anhydrous
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'