missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide, 99%, anhydrous

Product Code. 10388100 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10388100

Brand: Thermo Scientific Acros 207860250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13446-03-2 | Br2Mn | 214.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13446-03-2
Molecular Formula Br2Mn
Molecular Weight (g/mol) 214.76
MDL Number MFCD00011113

Specifications

Color Pink
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Linear Formula MnBr2
Merck Index 15, 5790
Solubility Information Solubility in water: soluble
Formula Weight 214.76
Percent Purity 99%
Packaging Glass bottle
Specific Gravity 4.38
Density 4.3800g/mL
Chemical Name or Material Manganese(II) bromide, Anhydrous
Show More Show Less
compliance-icons
Product Identifier
  • Manganese(II) bromide, anhydrous
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.