missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
32.00 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
Used as a desiccant, fertilizer and a catalyst. Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst. Used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 80°C |
Color | Pink |
Physical Form | Crystalline Aggregates |
Quantity | 5 g |
Linear Formula | Mn(OOCCH3)2·4H2O |
Merck Index | 14,5724 |
Solubility Information | Soluble in water,methanol,ethanol,and acetic acid. |
Formula Weight | 245.09 (173.03 Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'