missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis)

Product Code. 11396907
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396907 25 g 25g
11386907 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396907 Supplier Thermo Scientific Alfa Aesar Supplier No. 010802.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol

Used as a desiccant, fertilizer and a catalyst. Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst. Used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
IUPAC Name manganese(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80°C
Color Pink
Physical Form Crystalline Aggregates
Quantity 25 g
Linear Formula Mn(OOCCH3)2·4H2O
Merck Index 14,5724
Solubility Information Soluble in water,methanol,ethanol,and acetic acid.
Formula Weight 245.09 (173.03 Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Pungent
Density 1.589 g/mL
Assay (metals basis)
Chemical Name or Material Manganese(II) acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.