Learn More
Manganese(II) acetate tetrahydrate, Mn 22% (typical)
Description
It is used as a mordant in dyeing, drier for paints and varnishes, an oxidation catalyst, and in the manufacture of bister. It is also used in the manufacture of fertilizer. It is used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst
Specifications
Specifications
| Melting Point | 80°C |
| Color | Pink |
| Density | 1.589 g/mL |
| Odor | Pungent |
| Linear Formula | Mn(OOCCH3)2·4H2O |
| Quantity | 2 kg |
| Merck Index | 14,5724 |
| Solubility Information | Soluble in water,methanol,ethanol and acetic acid. |
| Formula Weight | 245.09 (173.03 Anhydrous) |
| Concentration or Composition (by Analyte or Components) | Mn 22% (typical) |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.