missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese(II) Acetate Tetrahydrate, ≥99.0% (KT), Honeywell™
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly Bottle
Description
- Products with a guarantee certificate and/or suitable for the stated analytical application.
Specifications
Melting Point | 80°C |
Color | Pink |
pH | Acidic |
Physical Form | Solid |
Quantity | 1 kg |
Linear Formula | (CH3COO)2Mn · 4H2O |
UN Number | NONH for all modes of transport |
Beilstein | 3730728 |
Formula Weight | 245.09g/mol |
Percent Purity | ≥99.0% (KT) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'