missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn

Product Code. 11378927
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378927 250 g 250g
11358927 1 kg 1kg
11368927 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378927 Supplier Thermo Scientific Alfa Aesar Supplier No. 038607.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13434-24-7 | C16H30MnO4 | 341.35 g/mol

Used as paint driers. Accelerating curing agent for unsaturated polyester. Also used as a intermediate for dyestuff, pharmaceutical, syntheses material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13434-24-7
Molecular Formula C16H30MnO4
Molecular Weight (g/mol) 341.35
MDL Number MFCD00058698
InChI Key FHRAKXJVEOBCBQ-UHFFFAOYSA-L
Synonym manganese bis 2-ethylhexanoate, manganese 2-ethylhexanoate, manganese 2+ bis 2-ethylhexanoate, hexanoic acid, 2-ethyl-, manganese 2+ salt, hexanoic acid, 2-ethyl-, manganese 2+ salt 2:1, hexanoic acid, 2-ethyl-, manganese salt, hexanoic acid, 2-ethyl-, manganese salt 1:?, manganese tallate 2-ethylhexanoate, manganous 2-ethylhexanoate, mmanganese 2-ethylhexanoate
PubChem CID 114521
IUPAC Name 2-ethylhexanoate;manganese(2+)
SMILES CCCCC(CC)C(=O)[O-].CCCCC(CC)C(=O)[O-].[Mn+2]

Specifications

Color Brown
Density 0.897 g/mL
Flash Point 40°C (104°F)
Odor Petroleum-like
Quantity 250 g
UN Number UN1268
Solubility Information Soluble in organic solvents. Not miscible or difficult to mix with water.
Formula Weight 341.35
Concentration or Composition (by Analyte or Components) 40% w/w in min.eral spirits, 6% Mn
Physical Form Liquid
Chemical Name or Material Manganese(II) 2-ethylhexanoate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Carcinogenicity Category 1B
  • Flammable liquid Category 3
  • Germ cell mutagenicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contain: Stoddard solvent

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.