missing translation for 'onlineSavingsMsg'
Learn More

Manganese granules, 0.8-12mm (0.03-0.47in), 99.98% (metals basis), Thermo Scientific Chemicals

Catalog Number 11363258
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
7439-96-5
Mn
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
23930
CHEBI:35154
manganese
[Mn]
This item is not returnable. View return policy
7439-96-5
Mn
MFCD00011111
PWHULOQIROXLJO-UHFFFAOYSA-N
colloidal, mangan, manganese, elemental, cutaval, magnacat, metal alloy, tronamang, fume, mangan polish, compounds
23930
CHEBI:35154
manganese
[Mn]

For vacuum deposition

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Assay Percent Range 99.98% (metals basis)
Quantity 25 g
Molecular Weight (g/mol) 54.94
Form Granular
Chemical Name or Material Manganese

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'