missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride Tetrahydrate, TRC

Product Code. 30478573
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30478573 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30478573

Brand: Toronto Research Chemicals TRCM16430010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Manganese Chloride (AS)
CAS 13446-34-9
Molecular Formula 2 Cl . Mn . 4 H2 O
Quantity 10 g
Synonym Manganese Chloride (MnCl2) Tetrahydrate, Bothdel Oral Solution 10, LumenHance, Manganese Chloride tetrahydrate, Manganese Dichloride Tetrahydrate, Manganese(2+) Chloride Tetrahydrate, Manganese(II) Chloride Tetrahydrate, Manganous Chloride Tetrahydrate
InChI Formula InChI=1S/2ClH.Mn.4H2O/h2*1H;;4*1H2/q;;+2;;;;/p-2
SMILES O.O.O.O.[Cl-].[Cl-].[Mn+2]
IUPAC Name manganese(2+);dichloride;tetrahydrate
Molecular Weight (g/mol) 197.91
Formula Weight 196.918
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.