missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride Tetrahydrate, (Crystalline),MP Biomedicals

Product Code. 11487612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487612 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11487612 Supplier MP Biomedicals Supplier No. 0215533490
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity crystalline manganese chloride used as a manganese ion source in various biological research applications.

Specifications

CAS 13446-34-9
Physical Form Crystalline
Quantity 500 g
Molecular Formula Cl2H8MnO4
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]
IUPAC Name manganese(2+) tetrahydrate dichloride
Molecular Weight (g/mol) 197.90
Formula Weight 197.9
Grade Research
Chemical Name or Material Manganese Chloride Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.