missing translation for 'onlineSavingsMsg'
Learn More

Manganese Carbonyl, TRC

Product Code. 30477190
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30477190 100 mg 100mg
30464643 10 mg 10mg
30480220 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30477190 Supplier Toronto Research Chemicals Supplier No. TRCM164250100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10170-69-1
Molecular Formula C10Mn2O10
Molecular Weight (g/mol) 389.98
Synonym Manganese Carbonyl, CORM 1, Decacarbonyldimanganese, Decacarbonyldimanganese(0), Dimanganese Decacarbonyl, Manganese Carbonyl (Mn2(CO)10), Manganese Pentacarbonyl Dimer, NSC 203018, NSC 22319
IUPAC Name carbon monoxide;manganese
SMILES [Mn].[Mn].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/10CO.2Mn/c10*1-2;;
Formula Weight 389.8252
Chemical Name or Material Manganese Carbonyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.