missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANF Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98366-50ul
This item is not returnable.
View return policy
Description
MANF Polyclonal antibody specifically detects MANF in Mouse samples. It is validated for ELISA
Specifications
| MANF | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Arginine-Rich Protein, arginine-rich, mutated in early stage tumors, ARMET, ARP, mesencephalic astrocyte-derived neurotrophic factor, Protein ARMET | |
| Produced in rabbits immunized with purified, recombinant Mouse MANF (Accession#: NP_083379.2; Met1-Leu179) | |
| 50 μg | |
| Biologically Active Proteins, Cancer, DNA Repair | |
| 7873 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction