missing translation for 'onlineSavingsMsg'
Learn More

MANF Antibody, DyLight 350, Novus Biologicals™

Product Code. 18430049 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430049 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430049 Supplier Novus Biologicals Supplier No. NBP299775UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MANF Polyclonal antibody specifically detects MANF in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen MANF
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Arginine-Rich Protein, arginine-rich, mutated in early stage tumors, ARMET, ARP, mesencephalic astrocyte-derived neurotrophic factor, Protein ARMET
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human MANF (Accession#: NP_006001.3; Met1-Leu182)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 7873
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.