missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MANEA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588029
This item is not returnable.
View return policy
Description
Immunogen sequence: RPNTATFGAP FGLDLLPELH QRTIHLGKNF DFQKSDRINS ETNTKNLKSV EITMKPSKAS ELNLDELPPL N; Positive Samples: 293T, U-87MG; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
N-glycosylation of proteins is initiated in the endoplasmic reticulum (ER) by the transfer of the preassembled oligosaccharide glucose-3-mannose-9-N-acetylglucosamine-2 from dolichyl pyrophosphate to acceptor sites on the target protein by an oligosaccharyltransferase complex. This core oligosaccharide is sequentially processed by several ER glycosidases and by an endomannosidase (E.C. 3.2.1.130), such as MANEA, in the Golgi. MANEA catalyzes the release of mono-, di-, and triglucosylmannose oligosaccharides by cleaving the alpha-1,2-mannosidic bond that links them to high-mannose glycans (Hamilton et al., 2005).
Specifications
| MANEA | |
| Polyclonal | |
| Unconjugated | |
| MANEA | |
| 4932703L02Rik; alpha 1,2-endomannosidase; ENDO; Endo-alpha mannosidase; endomannosidase; glycoprotein endo-alpha-1,2-mannosidase; hEndo; Mandaselin; Manea; mannosidase endo-alpha; mannosidase, endo-alpha; mEndo | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79694 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5SRI9 | |
| MANEA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 30-100 of human MANEA (NP_0789172). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction