missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANEA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£342.00 - £470.00
Specifications
| Antigen | MANEA |
|---|---|
| Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18435131
|
Novus Biologicals
NBP1-82279-25ul |
25 μL |
£342.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18207135
|
Novus Biologicals
NBP1-82279 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MANEA Polyclonal specifically detects MANEA in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MANEA | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| alpha 1,2-endomannosidase, DKFZp686D20120, EC 3.2.1, EC 3.2.1.130, ENDO, Endo-alpha mannosidase, endomannosidase, FLJ12838, glycoprotein endo-alpha-1,2-mannosidase, hEndo, mandaselin, mannosidase, endo-alpha | |
| MANEA | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 79694 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LRPNTATFGAPFGLDLLPELHQRTIHLGKNFDFQKSDRINSETNTKNLKSVEITMKPSKASELNLDELPPLNNYLHVFYYSWYGNPQFDGKYIHWNHPVLEHWDPRIAKNYPQGRHNPPDDIGSSFYPELGSYSSRD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title