missing translation for 'onlineSavingsMsg'
Learn More

Mandelic Acid, TRC

Product Code. 30009058
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009058 50 g 50g
30006292 10 g 10g
17924835 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009058 Supplier Toronto Research Chemicals Supplier No. TRCM16252550G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-64-2
Molecular Formula C8 H8 O3
Molecular Weight (g/mol) 152.15
Synonym (2RS)-2-Hydroxy-2-phenylacetic acid, Hydroxyphenylacetic acid, Mandelic acid, Glycopyrronium Bromide Imp. C (EP), Homatropine Methylbromide Imp. C (EP), Sertraline Hydrochloride Imp. E (EP) as Racemate, Benzeneacetic acid, α-hydroxy-, Mandelic acid (7CI,8CI), α-Hydroxybenzeneacetic acid, (RS)-Mandelic acid, (±)-2-Hydroxy-2-phenylethanoic acid, (±)-Mandelic acid, (±)-α-Hydroxybenzeneacetic acid, (±)-α-Hydroxyphenylacetic acid, 2-Hydroxy-2-phenylacetic acid, 2-Phenyl-2-hydroxyacetic acid, 2-Phenylglycolic acid, Almond acid, Amygdalic acid, DL-Amygdalic acid, DL-Hydroxy(phenyl)acetic acid, DL-Mandelic acid, NSC 7925, Paramandelic acid, Phenylglycolic acid, Phenylhydroxyacetic acid, S 14-75781, Uromaline, dl-Mandelic acid, α-Hydroxy-α-toluic acid, α-Hydroxyphenylacetic acid
IUPAC Name 2-hydroxy-2-phenylacetic acid
SMILES OC(C(=O)O)c1ccccc1

Specifications

InChI Formula InChI=1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)
Formula Weight 152.0473
Percent Purity >95%
Quantity 50 g
Chemical Name or Material DL-Mandelic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.