missing translation for 'onlineSavingsMsg'
Learn More

(±)-Mandelic Acid, 99%

Product Code. 11452327
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11452327 100 g 100g
11462327 500 g 500g
11472327 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452327 Supplier Thermo Scientific Alfa Aesar Supplier No. A14223.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 90-64-2 | C8H8O3 | 152.149 g/mol

Mandelic acid is used in the pharmaceutical industry as an antibacterial to treat urinary tract infections. It acts as a precursor for making various drugs. It is also used as an oral antibiotic. Mandelic acid peels are widely used for treating damaged skin due to its less penetrating and irritating property. Enantiomeric resolution of (±)-Mandelic acid is performed by using the chiral amine (1R,2S)-(-)-ephedrine followed by acidification to prepare (R)-( -)-Mandelic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mandelic acid is used in the pharmaceutical industry as an antibacterial to treat urinary tract infections. It acts as a precursor for making various drugs. It is also used as an oral antibiotic. Mandelic acid peels are widely used for treating damaged skin due to its less penetrating and irritating property. Enantiomeric resolution of (±)-Mandelic acid is performed by using the chiral amine (1R,2S)-(-)-ephedrine followed by acidification to prepare (R)-( -)-Mandelic acid.

Solubility
Soluble in water, diethyl ether, ethanol and isopropanol.

Notes
Hydrophobic and light sensitive. Incompatible with strong oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-64-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00064250
InChI Key IWYDHOAUDWTVEP-UHFFFAOYSA-N
Synonym mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid
PubChem CID 1292
ChEBI CHEBI:35825
IUPAC Name 2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

Melting Point 118°C to 121°C
Assay Percent Range 99%
Beilstein 510011
Merck Index 14,5717
Solubility Information Soluble in water,diethyl ether,ethanol and isopropanol.
Formula Weight 152.15
Percent Purity 99%
Quantity 100 g
Sensitivity Light sensitive
Chemical Name or Material (±)-Mandelic Acid
compliance-icons
Product Identifier
  • DL-Mandelic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: .alpha.-hydroxy-Benzeneacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.