missing translation for 'onlineSavingsMsg'
Learn More

Mandelic Acid ((2RS)-2-Hydroxy-2-phenylacetic Acid), LoGiCal

Product Code. 30458584
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458584 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458584 Supplier LoGiCal Supplier No. LGCFOR0792.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Mandelic Acid
CAS 90-64-2
Analyte or Component Names DL-Mandelic Acid
Shipping Condition Room Temperature
Molecular Formula C8 H8 O3
Certifications/Compliance ISO 9001
Synonym Glycopyrronium Bromide Imp. C (EP), Homatropine Methylbromide Imp. C (EP), Pregabalin Imp. C (EP), Sertraline Hydrochloride Imp. E (EP), Sertraline Imp. E (EP), (2RS)-2-Hydroxy-2-phenylacetic Acid, Hydroxyphenylacetic Acid, Mandelic Acid, Sertraline Hydrochloride Imp. E (EP) as Racemate, Mandelic acid, (RS)-2-Hydroxy-2-phenylacetic acid, (RS)-Mandelic acid, (+/-)-2-Hydroxy-2-phenylethanoic acid, (+/-)-Mandelic acid, (+/-)-alpha-Hydroxybenzeneacetic acid, (+/-)-alpha-Hydroxyphenylacetic acid, 2-Hydroxy-2-phenylacetic acid, 2-Phenyl-2-hydroxyacetic acid, 2-Phenylglycolic acid, Almond acid, Amygdalic acid, DL-Amygdalic acid, DL-Hydroxy(phenyl)acetic acid, DL-Mandelic acid, NSC 7925, Paramandelic acid, Phenylglycolic acid, Phenylhydroxyacetic acid, Uromaline, dl-Mandelic acid, alpha-Hydroxy-alpha-toluic acid, alpha-Hydroxybenzeneacetic acid, alpha-Hydroxyphenylacetic acid, Glycopyrronium Bromide Impurity C, Homatropine Methylbromide Impurity C, Pregabalin Impurity C, Sertraline Hydrochloride Impurity E as Racemate, Glycopyrronium Impurity C, Sertraline Impurity E as Racemate
InChI Formula InChI=1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)
Solution Type Neat
SMILES OC(C(=O)O)c1ccccc1
IUPAC Name 2-hydroxy-2-phenylacetic acid
Industry Type Clinical Testing
Molecular Weight (g/mol) 152.15
Quantity 10 mg
Formula Weight 152.0473 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.