missing translation for 'onlineSavingsMsg'
Learn More

Mancozeb (Technical Grade), TRC

Product Code. 30474103
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30474103 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30474103 Supplier Toronto Research Chemicals Supplier No. TRCM162200500MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Mancozeb
CAS 8018-01-7
Molecular Formula C4 H6 Mn N2 S4 . C4 H6 N2 S4 Zn
Quantity 500 mg
Synonym Manganese, [N-[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']-, mixt. with [N-[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']zinc (ACI), Carbamic acid, ethylenebis[dithio-, manganese zinc complex (8CI), Carbamodithioic acid, 1,2-ethanediylbis-, manganese complex (ZCI), Carbamodithioic acid, 1,2-ethanediylbis-, zinc complex (ZCI), Manganese, [[1,2-ethanediylbis[carbamodithioato]](2-)]-, mixt. with [[1,2-ethanediylbis[carbamodithioato]](2-)]zinc (ZCI), Manganese, [[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']-, mixt. with [[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']zinc (9CI), Zinc, [[1,2-ethanediylbis[carbamodithioato]](2-)]-, mixt. contg. (ZCI), Zinc, [[2-[(dithiocarboxy)amino]ethyl]carbamodithioato(2-)-κS,κS']-, mixt. contg. (9CI), Agrox 16D, Agrozeb 80WP, Banazeb 60SC, Carmazine, Contizeb, Daisenmengxin, Dhanuka M 45, Diesen M, Dithane, Dithane 60SC, Dithane 945, Dithane DG, Dithane DG NT, Dithane F, Dithane F 45, Dithane M 45, Dithane M45, Dithane M80, Dithane Neo Tec 75WG, Dithane NT, Dithane OS, Dithane Rainshield Neo Tec, Dithane S 60, Dithane SPC, Dithane ultra, F 2966, Fore, Green-daisen M, Greenzeb 80WP, Hi-shield, Indofil 80WP, Indofil M 45, Indofil M45, Jimanaisen, Jimandaisen, Karamate, Karamate N, Kascade, Liro manzeb, Mancomix, Mancosan, Mancozan, Mancozeb, Mancozebe Sipcam, Mancozi, Maneb-zinc, Mankogal, Manoseb, Manzate 200, Manzeb, Manzidan, Marzin, Milcozebe, Mount 45, Mycotrin, Nemispor, Novozir MN, Novozir MN 80, Othane M 45, Pencozeb, Penncozeb, Pennfluid, Sakozeb Blue, Sancozeb, Sandozeb Pepite, Suncozeb, Tanzeo M 45, Thane M 45, Trimanoc, Tritogol MZ, Vondozeb, Ziman-Dithane, Zimanat, Zimaneb, Zinc manganese ethylenebisdithiocarbamate
InChI Formula InChI=1S/2C4H8N2S4.Mn.Zn/c2*7-3(8)5-1-2-6-4(9)10;;/h2*1-2H2,(H2,5,7,8)(H2,6,9,10);;/q;;2*+2/p-4
SMILES S=C1NCCNC(=S)S[Zn]S1.S=C2NCCNC(=S)S[Mn]S2
IUPAC Name 1,3-dithia-5,8-diaza-2λ^{2}-manganacyclononane-4,9-dithione;1,3-dithia-5,8-diaza-2λ^{2}-zincacyclononane-4,9-dithione
Molecular Weight (g/mol) 541.07
Formula Weight 538.75
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.