missing translation for 'onlineSavingsMsg'
Learn More

Mancozeb, MedChemExpress

Product Code. 16452289
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16452289 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452289 Supplier Medchem Express Supplier No. HYB0854
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mancozeb is an ethylene-bis-dithiocarbamate fungicide.

Mancozeb is an ethylene-bis-dithiocarbamate fungicide.

Specifications

Chemical Name or Material Mancozeb
Physical Form Solid
CAS 8018-01-7
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C8H12Mn2N4S8Zn2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 1 mg/mL (1.51 mM; ultrasonic and warming and heat to 80°C) ∣DMSO : 1 mg/mL (1.51 mM; Need ultrasonic)
SMILES S=C([S-])NCCNC1=[S][Mn]([Mn+])[S-]1.S=C([S-])NCCNC2=[S][Zn]([Zn+])[S-]2
Molecular Weight (g/mol) 661.36
Formula Weight 661.36
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.