missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANBAL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16028-1-AP-20UL
This item is not returnable.
View return policy
Description
MANBAL is a protein coding gene.Specifications
| MANBAL | |
| Polyclonal | |
| Unconjugated | |
| MANBAL | |
| MANBAL, Protein MANBAL | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 499934, 63905, 69161 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9D8X0, Q9NQG1 | |
| MANBAL | |
| MANBAL Fusion Protein Ag8888 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction