missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANBA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32355
This item is not returnable.
View return policy
Description
MANBA Polyclonal specifically detects MANBA in Human samples. It is validated for Western Blot.
Specifications
| MANBA | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| MANBA | |
| Recombinant protein encompassing a sequence within the center region of human MANBA. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 4126 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| beta-mannosidase, EC 3.2.1.25, Lysosomal beta A mannosidase, MANB1mannanase, Mannanase, Mannase, mannosidase, beta A, lysosomal | |
| Rabbit | |
| 101 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction