missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MANBA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99160-50ul
This item is not returnable.
View return policy
Description
MANBA Polyclonal antibody specifically detects MANBA in Human samples. It is validated for Western Blot
Specifications
| MANBA | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| beta-mannosidase, EC 3.2.1.25, Lysosomal beta A mannosidase, MANB1mannanase, Mannanase, Mannase, mannosidase, beta A, lysosomal | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MANBA | |
| 50 μg | |
| Cancer, Endocrinology, Neuroscience, Signal Transduction | |
| 4126 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction