missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAN2A1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518851
This item is not returnable.
View return policy
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a glycosyl hydrolase that localizes to the Golgi and catalyzes the final hydrolytic step in the asparagine-linked oligosaccharide (N-glycan) maturation pathway. Mutations in the mouse homolog of this gene have been shown to cause a systemic autoimmune disease similar to human systemic lupus erythematosus.
Specifications
| MAN2A1 | |
| Polyclonal | |
| Unconjugated | |
| MAN2A1 | |
| Alpha-mannosidase 2; alpha-mannosidase II; AMan II; Golgi alpha-mannosidase II; golgi integral membrane protein 7; GOLIM7; man II; Man2a1; Mana2; Mana-2; MANII; Manno; mannosidase 2, alpha 1; mannosidase alpha 2; mannosidase alpha class 2A member 1; mannosidase, alpha type II; mannosidase, alpha, class 2A, member 1; mannosyl-oligosaccharide 1,3-1,6-alpha-mannosidase; Map-2 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 4124 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q16706 | |
| MAN2A1 | |
| Synthetic peptide sequence (KISSDIKSQNR) corresponding to the internal amino acids of MAN2A1 (aa 881-892). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction