missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN1C1 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£151.00 - £366.00
Specifications
| Antigen | MAN1C1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18361977
|
Novus Biologicals
NBP3-05040-20UL |
20 μL |
£151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18374535
|
Novus Biologicals
NBP3-05040-100UL |
100 μL |
£366.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAN1C1 Polyclonal specifically detects MAN1C1 in Human, Mouse samples. It is validated for Western Blot.Specifications
| MAN1C1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 1,2-alpha-mannosidase IC, Alpha-1,2-mannosidase IC, EC 3.2.1.113, HMICProcessing alpha-1,2-mannosidase IC, MAN1A3, MAN1C, Mannosidase alpha class 1C member 1, mannosidase, alpha, class 1C, member 1, mannosyl-oligosaccharide 1,2-alpha-mannosidase IC, pp6318 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 44-240 of human MAN1C1 (NP_065112.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS with 50% glycerol, pH7.3. | |
| 57134 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title