missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: QSQVRAQREK IKEMMQFAWQ SYKRYAMGKN ELRPLTKDGY EGNMFGGLSG ATVIDSLDTL YLMELKEEFQ EAKAWVGESF HLNVSG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
MAN1C1 gene ontology annotations related to this gene include metabolic process.
Specifications
Specifications
| Antigen | MAN1C1 |
| Applications | Immunohistochemistry |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | MAN1C1 |
| Gene Accession No. | Q9NR34 |
| Gene Alias | 1,2-alpha-mannosidase IC; AI593348; Alpha-1,2-mannosidase IC; HMIC; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; MAN1A3; MAN1C; MAN1C1; Mannosidase alpha class 1C member 1; mannosidase, alpha, class 1C, member 1; mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; pp6318; processing alpha-1,2-mannosidase IC |
| Gene Symbols | MAN1C1 |
| Show More |
Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?