missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAN1B1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104494
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MAN1B1.
Involved in glycoprotein quality control targeting of misfolded glycoproteins for degradation. It primarily trims a single alpha-1,2-linked mannose residue from Man9GlcNAc2 to produce Man8GlcNAc2, but at high enzyme concentrations, as found in the ER quality control compartment (ERQC), it further trims the carbohydrates to Man5-6GlcNAc2. Tremblay L.O., Glycobiology 9:1073-1078(1999) [PubMed: 10521544]. Gonzalez D.S., J. Biol. Chem. 274:21375-21386(1999) [PubMed: 10409699]. Clark H.F., Genome Res. 13:2265-2270(2003) [PubMed: 12975309].
Specifications
| MAN1B1 | |
| Polyclonal | |
| Unconjugated | |
| MAN1B1 | |
| 3.2.1.113; E430019H13Rik; endoplasmic Reticulum Class I alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase; endoplasmic reticulum mannosyl-oligosaccharide 1,2-alpha-mannosidase 1; ER alpha 1,2-mannosidase; ER alpha-1,2-mannosidase; ER mannosidase 1; ERMan1; ERManI; Gm108; MAN1B1; Man9GlcNAc2-specific processing alpha-mannosidase; man9GlcNAc2-specific-processing alpha-mannosidase; MANA-ER; Mannosidase alpha class 1B member 1; mannosidase, alpha, class 1B, member 1; MRT15; RGD1563595; RP11-350O14.2; UNQ747/PRO1477 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11253, 227619, 499751 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A2AJ15, B2GUY0, Q9UKM7 | |
| MAN1B1 | |
| A synthesized peptide derived from human MAN1B1(Accession Q9UKM7), corresponding to amino acid residues V135-A185. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction