missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN1B1, Mouse anti-Human, Clone: 6B1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00011253-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: AFRLEEEQKMRPEIAGLKPANPPVLPAPQKADTDPENLPEISSQKTQRHIQRGPPHLQIRPPSQDLKDGTQEEATKRQEAPVDPRPEGDPQRTVISWRGASpecifications
| MAN1B1 | |
| Monoclonal | |
| Unconjugated | |
| mannosidase, alpha, class 1B, member 1 | |
| MANA-ER | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 6B1 | |
| In 1x PBS, pH 7.4 | |
| NM_016219 | |
| MAN1B1 | |
| MAN1B1 (NP_057303, 112 a.a. ∼ 211 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 11253 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction