missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAN1A2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35426-100ul
This item is not returnable.
View return policy
Description
MAN1A2 Polyclonal antibody specifically detects MAN1A2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MAN1A2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Alpha-1,2-mannosidase IB, EC 3.2.1.113, MAN1Balpha1,2-mannosidase, Mannosidase alpha class 1A member 2, mannosidase, alpha, class 1A, member 2, mannosyl-oligosaccharide 1,2-alpha-mannosidase IB, Processing alpha-1,2-mannosidase IB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-180 of human MAN1A2 (NP_006690.1).,, Sequence:, SSKHKRFDLGLEDVLIPHVDAGKGAKNPGVFLIHGPDEHRHREEEERLRNKIRADHEKALEEAKEKLRKSREEIRAEIQTEKNKVVQEMKIKENKPLPPVPIPNLVGIRGGDPEDNDIREK | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10905 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction