Learn More
Emma Safety Footwear Mammoet M1966 D Safety Shoes
State-of-the art Sympatex™ construction provides perfect foot protection for 'hands on' roles within the Chemical Industry and Agriculture. Virtually waterproof, these boots allow feet to breathe properly, keeping them cool and dry.
Brand:  Emma Safety Footwear 584848-49
Additional Details : Weight : 2.67000kg
 View more versions of this product
Product Code. 15978811
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Duo Heat Resistant Outsole
  • Sympatex™ Waterproof
  • Heat and cold insulated
  • Sanitized
Specifications
Specifications
EN ISO 20345:2011
Mammoet M1966 D
Oiled Nubuck
Steel
Black
Hydro-Tec™ Sanitized Silver / Sympatex™
49