missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Mouse anti-Human, Clone: MGB/4056, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | Mammaglobin A |
|---|---|
| Clone | MGB/4056 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
Mammaglobin A Monoclonal specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| Mammaglobin A | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| Recombinant full-length human Mammaglobin A protein | |
| Primary | |
| Store at 4°C. |
| MGB/4056 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 4250 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title