missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Antibody (MGB/7980R), Novus Biologicals™
Rabbit Monoclonal Antibody
£224.00 - £412.00
Specifications
| Antigen | Mammaglobin A |
|---|---|
| Clone | MGB/7980R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650915
|
Novus Biologicals
NBP3-20441-20ug |
20 μg |
£224.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18610344
|
Novus Biologicals
NBP3-20441-100ug |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Mammaglobin A | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| Recombinant fragment of human Mammaglobin A protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
| MGB/7980R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10mM PBS with 0.05% BSA | |
| 4250 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title