missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mammaglobin A Antibody (CL8892) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05504B
This item is not returnable.
View return policy
Description
Mammaglobin A Monoclonal antibody specifically detects Mammaglobin A in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| Mammaglobin A | |
| Monoclonal | |
| Biotin | |
| mammaglobin 1, mammaglobin A, Mammaglobin-1, MGB1mammaglobin-A, MGC71974, Secretoglobin family 2A member 2, secretoglobin, family 2A, member 2, UGB2mammaglobin-1 | |
| This antibody was developed using a synthetic peptide derived from Q13296, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL8892 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 4250 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction