missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAMLD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552025
This item is not returnable.
View return policy
Description
Immunogen sequence: KRPCLEDVTL AMGPGAHPST ACAELQVPPL TINPSPAAMG VAGQSLLLEN NPMNGNIMGS PFVVPQTTEV GLKGPTVPYY EKINSVPAVD QELQELLEEL TKIQDPSPNE LDLE Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 25%.
CXorf6 is a protein predicted based on an ORF found in chromosome 14.
Specifications
| MAMLD1 | |
| Polyclonal | |
| Unconjugated | |
| MAMLD1 | |
| CG1; CXorf6; F18; G630014P10Rik; HYSP2; MAMLD1; mastermind like domain containing 1; mastermind-like domain containing 1; mastermind-like domain-containing protein 1; Protein CG1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10046 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13495 | |
| MAMLD1 | |
| Recombinant protein corresponding to Human MAMLD1. Recombinant protein control fragment (Product #RP-91982). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction