missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAML2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568932
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 79%; Horse: 92%; Human: 92%; Mouse: 100%; Rabbit: 92%; Rat: 93%.
Acts as a transcriptional coactivator for NOTCH proteins. Has been shown to amplify NOTCH-induced transcription of HES1. Potentiates activation by NOTCH3 and NOTCH4 more efficiently than MAML1 or MAML3.
Specifications
| MAML2 | |
| Polyclonal | |
| Unconjugated | |
| MAML2 | |
| 5930431H10; BC032967; KIAA1819; MAM2; Mam-2; MAM3; MAM-3; MAML2; mastermind like 2 (Drosophila); mastermind like transcriptional coactivator 2; mastermind-like 2; mastermind-like protein 2; mastermind-like transcriptional coactivator 2; MLL-MAML2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 270118 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| MAML2 | |
| synthetic peptide directed towards the C-terminal region of mouse MAML2. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering