missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAML1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55493-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to MAML1. The calculated molecular weight of MAML1 is 108 kDa, but modified MAML1 is about 120 kDa.
This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway.Specifications
| MAML1 | |
| Polyclonal | |
| Unconjugated | |
| MAML1 | |
| KIAA0200, Mam 1, Mam1, MAML1, mastermind like 1 (Drosophila), Mastermind like protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 103806, 303101, 9794 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.38 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6T264, Q92585 | |
| Maml1 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction