missing translation for 'onlineSavingsMsg'
Learn More

MAML1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55493-1-AP-150UL

 View more versions of this product

Product Code. 16826955

  • £349.00 / 150µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is specific to MAML1. The calculated molecular weight of MAML1 is 108 kDa, but modified MAML1 is about 120 kDa.

This protein is the human homolog of mastermind, a Drosophila protein that plays a role in the Notch signaling pathway involved in cell-fate determination. There is in vitro evidence that the human homolog forms a complex with the intracellular portion of human Notch receptors and can increase expression of a Notch-induced gene. This evidence supports its proposed function as a transcriptional co-activator in the Notch signaling pathway.
Specifications

Specifications

MAML1
Polyclonal
Unconjugated
MAML1
KIAA0200, Mam 1, Mam1, MAML1, mastermind like 1 (Drosophila), Mastermind like protein 1
Rabbit
Antigen Affinity Chromatography
RUO
103806, 303101, 9794
-20°C
Liquid
Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6T264, Q92585
Maml1
Peptide
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.