missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAML1 Polyclonal Antibody

MAML1 Polyclonal Antibody
Brand: Invitrogen PA595871
This item is not returnable.
View return policy
Description
MAML1 Polyclonal Antibody for Western Blot
Specifications
| MAML1 | |
| Polyclonal | |
| Unconjugated | |
| MAML1 | |
| AI644666; AW743257; D930008C07Rik; FLJ53540; KIAA0200; Mam1; Mam-1; MAML1; mastermind homolog; mastermind like 1 (Drosophila); mastermind like transcriptional coactivator 1; mastermind-like 1; mastermind-like protein 1; mastermind-like transcriptional coactivator 1; mKIAA0200 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 103806, 303101, 9794 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.14 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q6T264, Q92585 | |
| MAML1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human MAML1 (NP_0555721). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction