missing translation for 'onlineSavingsMsg'
Learn More

Maltotriose, MedChemExpress

Product Code. 16405566
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16405566 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16405566 Supplier Medchem Express Supplier No. HY113011
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Maltotriose, the second most abundant sugar present in brewing, is an inducer of the maltose regulon of Escherichia coli. Maltotriose can induce beta-galactosidase synthesis.

Maltotriose, the second most abundant sugar present in brewing, is an inducer of the maltose regulon of Escherichia coli. Maltotriose can induce beta-galactosidase synthesis.

Specifications

Chemical Name or Material Maltotriose
Physical Form Solid
CAS 1109-28-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H32O16
Solubility Information H2O : 250 mg/mL (495.60 mM; Need ultrasonic)
SMILES OC[C@@H](O[C@@]1(O[C@@H]2[C@@H](CO)O[C@@H]([C@@H]([C@H]2O)O)O[C@@]([H])([C@@H]([C@H](C=O)O)O)[C@@H](CO)O)[H])[C@H]([C@@H]([C@H]1O)O)O
Molecular Weight (g/mol) 504.44
Formula Weight 504.44
Percent Purity 96.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.