missing translation for 'onlineSavingsMsg'
Learn More

Maltotriose, 93%, ACROS Organics™

Brand:  Acros Organics 225940250

Code : 29

Additional Details : CAS Number : 1109-28-0 Weight : 0.08000kg

Packaging Glass bottle

 View more versions of this product

Product Code. 10127843

  • £433.30 / 25g


EU Stock 4
Add to basket

Specifications

Chemical Identifiers

Maltotriose
93%
C18H32O16
MFCD00006629
1109-28-0
93%
504.44

Specifications

Maltotriose
White
25g
Fine Crystalline Powder or Crystals
93%
Authentic
MFCD00006629
+ 162.50
+162.50° (25°C c=1,H2O)
132.0° to 135.0°C
1109-28-0
93%
C18H32O16
504.44
Glass bottle
5% max. (K.F.)
Videos
SDS