missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltose Binding Protein Rabbit anti-E. coli, Clone: S02-6F4, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in E. coli samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Maltose Binding Protein |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S02-6F4 |
| Conjugate | Unconjugated |
| Formulation | 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide |
| Gene Alias | Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human Maltose Binding Protein (Uniprot#: P0AEX9) |
| Purification Method | Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?