missing translation for 'onlineSavingsMsg'
Learn More

Maltose Binding Protein Mouse anti-E. coli, Clone: R29.6, Novus Biologicals™

Mouse Monoclonal Antibody

£405.00

Specifications

Antigen Maltose Binding Protein
Clone R29.6
Concentration 0.2 mg/mL
Dilution Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL
Applications Western Blot, Immunofluorescence
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18605736
View Documents
Novus Biologicals
NBP3-07585-100ug
100 μg
£405.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in E. coli samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Maltose Binding Protein
0.2 mg/mL
Western Blot, Immunofluorescence
Unconjugated
Mouse
E. coli
Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
IgG1 κ
Protein A or G purified
R29.6
Western Blot 1 to 2 μg/mL, Immunocytochemistry/Immunofluorescence 1 to 2 μg/mL
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
MOS maltose binding protein fusion protein.
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.