missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltose Binding Protein Antibody (TVG MBP 3), Novus Biologicals™
Mouse Monoclonal Antibody
£149.00 - £316.00
Specifications
| Antigen | Maltose Binding Protein |
|---|---|
| Clone | TVG MBP 3 |
| Dilution | Western Blot : 1:100 - 1:2000, ELISA, Immunoprecipitation : 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624819
|
Novus Biologicals
NBP2-76653-0.025ml |
0.025 mL |
£149.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18352290
|
Novus Biologicals
NBP2-76653 |
0.1 mL |
£316.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, ELISA, Immunoprecipitation.Specifications
| Maltose Binding Protein | |
| Western Blot : 1:100 - 1:2000, ELISA, Immunoprecipitation : 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| Non-species specific | |
| Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein | |
| IgM | |
| Thiophilic Chromatography | |
| This antibody is useful for detection and isolation of recombinant MBP fusion proteins. |
| TVG MBP 3 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| PBS with 0.02% sodium azide | |
| A single intravenous injection of the maltose binding protein MBP-E2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title