missing translation for 'onlineSavingsMsg'
Learn More

Maltose Binding Protein Antibody (R3.2), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18414358 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414358 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414358 Supplier Novus Biologicals Supplier No. NBP276652MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Maltose Binding Protein
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone R3.2
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
Host Species Mouse
Immunogen MOS maltose binding protein fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Epitope Tags, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.