missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maltose Binding Protein Antibody (R29.6), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Maltose Binding Protein Monoclonal antibody specifically detects Maltose Binding Protein in Nonspecies specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP)
Specifications
Specifications
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) |
| Gene Alias | Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein |
| Host Species | Mouse |
| Form | Purified |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?