missing translation for 'onlineSavingsMsg'
Learn More

Maltose Binding Protein Antibody (R29.6), DyLight 405, Novus Biologicals™

Product Code. 18327430 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327430 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327430 Supplier Novus Biologicals Supplier No. NBP276651V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Maltose Binding Protein Monoclonal specifically detects Maltose Binding Protein in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Maltose Binding Protein
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone R29.6
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Mal E, MalE, Maltodextrin binding protein, Maltose ABC transporter periplasmic protein, Maltose binding periplasmic protein, MBP, MMBP, Periplasmic maltose binding protein
Host Species Mouse
Immunogen MOS maltose binding protein fusion protein
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity This antibody is useful for detection and isolation of recombinant MBP fusion proteins.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.